Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCATGTATTGGTTACCATACTA[A/G]GTACTTTGCATAGATGATTGTATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
VWA3A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
VWA3A - von Willebrand factor A domain containing 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173615.4 | Intron | NP_775886.3 | ||||
XM_011545742.2 | Intron | XP_011544044.1 | ||||
XM_011545744.2 | Intron | XP_011544046.1 | ||||
XM_011545745.1 | Intron | XP_011544047.1 | ||||
XM_011545746.2 | Intron | XP_011544048.1 | ||||
XM_017022947.1 | Intron | XP_016878436.1 | ||||
XM_017022948.1 | Intron | XP_016878437.1 | ||||
XM_017022949.1 | Intron | XP_016878438.1 |