Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCGCAAGCTCGCACTCATGAGTC[A/G]GAGAGACAGGGCCGTGAGCCCCAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604375 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARL16 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARL16 - ADP ribosylation factor like GTPase 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HGS - hepatocyte growth factor-regulated tyrosine kinase substrate | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004712.4 | Intron | NP_004703.1 | ||||
XM_011525463.2 | Intron | XP_011523765.1 | ||||
XM_017025297.1 | Intron | XP_016880786.1 | ||||
XM_017025298.1 | Intron | XP_016880787.1 |
MIR6786 - microRNA 6786 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |