Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCAAGTCCTAGTTCACTCAGGTT[C/T]TGTTAAGTTAGCAGTTGAAAATAAT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
|||||||||||||||||||||||
Phenotype: |
MIM: 603809 | |||||||||||||||||||||||
Literature Links: |
THRAP3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
THRAP3 - thyroid hormone receptor associated protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321471.1 | Intron | NP_001308400.1 | ||||
NM_001321473.1 | Intron | NP_001308402.1 | ||||
NM_005119.3 | Intron | NP_005110.2 | ||||
XM_005271371.4 | Intron | XP_005271428.1 |
Set Membership: |
HapMap |