Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTTTGCAACCTCAAAGATGGTTT[G/T]AGTTTTAGTTCCCATGAGTACATCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
43 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601883 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf174 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C1orf174 - chromosome 1 open reading frame 174 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207356.2 | 3516 | UTR 3 | NP_997239.2 | |||
XM_005244743.3 | 3516 | UTR 3 | XP_005244800.1 | |||
XM_011541323.2 | 3516 | UTR 3 | XP_011539625.1 |
DFFB - DNA fragmentation factor subunit beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |