Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGAAGCTAAATTTTTGAGTCCTT[A/G]CTTGTCTAAAAATGGCTTTATTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609017 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERMAP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERMAP - erythroblast membrane associated protein (Scianna blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017922.1 | Intron | NP_001017922.1 | ||||
NM_018538.3 | Intron | NP_061008.2 | ||||
XM_006710313.3 | Intron | XP_006710376.1 | ||||
XM_011540570.2 | Intron | XP_011538872.1 | ||||
XM_011540571.2 | Intron | XP_011538873.1 |
SVBP - small vasohibin binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |