Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGAAGCGATCTCATCGAAGGTCC[G/T]GCCTTTAGTCTCAGGAACTTTGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
26 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138140 MIM: 143090 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC2A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLC2A1 - solute carrier family 2 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006516.2 | 1897 | Silent Mutation | NP_006507.2 |