Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAGAGGCAGAAGAGTCAGGGTCA[C/G]AGGAGGAGCCATCATGATAGGAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC129 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC129 - coiled-coil domain containing 129 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257967.1 | Intron | NP_001244896.1 | ||||
NM_001257968.1 | Intron | NP_001244897.1 | ||||
NM_194300.3 | Intron | NP_919276.2 | ||||
XM_011515213.1 | Intron | XP_011513515.1 | ||||
XM_017011871.1 | Intron | XP_016867360.1 | ||||
XM_017011872.1 | Intron | XP_016867361.1 |