Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602308 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
WWP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
WWP2 - WW domain containing E3 ubiquitin protein ligase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270453.1 | Intron | NP_001257382.1 | ||||
NM_001270454.1 | Intron | NP_001257383.1 | ||||
NM_001270455.1 | Intron | NP_001257384.1 | ||||
NM_007014.4 | Intron | NP_008945.2 | ||||
NM_199424.2 | Intron | NP_955456.1 | ||||
XM_011522823.2 | Intron | XP_011521125.1 | ||||
XM_011522825.1 | Intron | XP_011521127.1 | ||||
XM_011522826.2 | Intron | XP_011521128.1 | ||||
XM_017022879.1 | Intron | XP_016878368.1 | ||||
XM_017022880.1 | Intron | XP_016878369.1 | ||||
XM_017022881.1 | Intron | XP_016878370.1 |
Set Membership: |
HapMap |