Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTTTTATTTATATTGTCACTTTT[C/T]GCCAATTAGAAATTTTAATAGATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606686 MIM: 601750 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF2B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EIF2B1 - eukaryotic translation initiation factor 2B subunit alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GTF2H3 - general transcription factor IIH subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271866.1 | Intron | NP_001258795.1 | ||||
NM_001271867.1 | Intron | NP_001258796.1 | ||||
NM_001271868.1 | Intron | NP_001258797.1 | ||||
NM_001516.4 | Intron | NP_001507.2 | ||||
XM_017019228.1 | Intron | XP_016874717.1 |