Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605615 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARIH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARIH2 - ariadne RBR E3 ubiquitin protein ligase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317333.1 | Intron | NP_001304262.1 | ||||
NM_001317334.1 | Intron | NP_001304263.1 | ||||
NM_006321.3 | Intron | NP_006312.1 | ||||
XM_011533265.1 | Intron | XP_011531567.1 | ||||
XM_011533266.1 | Intron | XP_011531568.1 | ||||
XM_011533267.1 | Intron | XP_011531569.1 | ||||
XM_011533268.1 | Intron | XP_011531570.1 | ||||
XM_011533270.1 | Intron | XP_011531572.1 | ||||
XM_011533271.1 | Intron | XP_011531573.1 | ||||
XM_011533273.1 | Intron | XP_011531575.1 | ||||
XM_011533275.2 | Intron | XP_011531577.1 | ||||
XM_011533276.1 | Intron | XP_011531578.1 | ||||
XM_017005531.1 | Intron | XP_016861020.1 | ||||
XM_017005532.1 | Intron | XP_016861021.1 | ||||
XM_017005533.1 | Intron | XP_016861022.1 | ||||
XM_017005534.1 | Intron | XP_016861023.1 | ||||
XM_017005535.1 | Intron | XP_016861024.1 | ||||
XM_017005536.1 | Intron | XP_016861025.1 | ||||
XM_017005537.1 | Intron | XP_016861026.1 | ||||
XM_017005538.1 | Intron | XP_016861027.1 | ||||
XM_017005539.1 | Intron | XP_016861028.1 | ||||
XM_017005540.1 | Intron | XP_016861029.1 | ||||
XM_017005541.1 | Intron | XP_016861030.1 |
ARIH2OS - ariadne homolog 2 opposite strand | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |