Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCCCTTTTAGCAGATAAAGGAGG[G/T]GGCATTTGTGGAAGGAGGTCCTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608250 | ||||||||||||||||||||
Literature Links: |
SUDS3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SUDS3 - SDS3 homolog, SIN3A corepressor complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022491.2 | Intron | NP_071936.2 | ||||
XM_011538671.2 | Intron | XP_011536973.1 | ||||
XM_017019817.1 | Intron | XP_016875306.1 | ||||
XM_017019818.1 | Intron | XP_016875307.1 | ||||
XM_017019819.1 | Intron | XP_016875308.1 | ||||
XM_017019820.1 | Intron | XP_016875309.1 |