Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGCTGACTAGGTTATCCGCAGG[A/G]GGTAACATATTATGGTGAGAGGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM19A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FAM19A2 - family with sequence similarity 19 member A2, C-C motif chemokine like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178539.4 | Intron | NP_848634.1 | ||||
XM_006719370.2 | Intron | XP_006719433.1 | ||||
XM_011538272.1 | Intron | XP_011536574.1 | ||||
XM_011538274.1 | Intron | XP_011536576.1 |
Set Membership: |
HapMap |