Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACTAGCTGAAGCACAAAGACAGA[A/G]ACATCTAATTCTTATTTGTGTTACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612604 MIM: 612605 MIM: 612606 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LCE1B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LCE1B - late cornified envelope 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LCE1C - late cornified envelope 1C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276331.1 | Intron | NP_001263260.1 | ||||
NM_178351.3 | Intron | NP_848128.1 |
LCE1D - late cornified envelope 1D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |