Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCACTTATTGAAAAACGGGCTTT[C/T]AAAAACAACTTACTGAGGTATAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610894 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NRG4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NRG4 - neuregulin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138573.3 | Intron | NP_612640.1 | ||||
XM_017021937.1 | Intron | XP_016877426.1 | ||||
XM_017021938.1 | Intron | XP_016877427.1 | ||||
XM_017021939.1 | Intron | XP_016877428.1 | ||||
XM_017021940.1 | Intron | XP_016877429.1 | ||||
XM_017021941.1 | Intron | XP_016877430.1 | ||||
XM_017021942.1 | Intron | XP_016877431.1 | ||||
XM_017021943.1 | Intron | XP_016877432.1 | ||||
XM_017021944.1 | Intron | XP_016877433.1 | ||||
XM_017021945.1 | Intron | XP_016877434.1 | ||||
XM_017021946.1 | Intron | XP_016877435.1 | ||||
XM_017021947.1 | Intron | XP_016877436.1 | ||||
XM_017021948.1 | Intron | XP_016877437.1 |
Set Membership: |
HapMap |