Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAATATGCACACACATTTACTTA[A/G]GGCAGATATTTAGTTACCTAGGGCA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 300724 | |||||||||||||||||||||||
Literature Links: |
CNKSR2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
CNKSR2 - connector enhancer of kinase suppressor of Ras 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168647.1 | Intron | NP_001162118.1 | ||||
NM_001168648.1 | Intron | NP_001162119.1 | ||||
NM_001168649.1 | Intron | NP_001162120.1 | ||||
NM_014927.3 | Intron | NP_055742.2 | ||||
XM_011545471.2 | Intron | XP_011543773.1 | ||||
XM_011545472.2 | Intron | XP_011543774.1 | ||||
XM_017029358.1 | Intron | XP_016884847.1 |