Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTATTAAGTACATCAAAATTTC[A/C]CCTTACATTTGAAATTGCATAGAAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 605905 | |||||||||||||||||||||||
Literature Links: |
DCUN1D1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
DCUN1D1 - defective in cullin neddylation 1 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308101.1 | 3615 | UTR 3 | NP_001295030.1 | |||
NM_020640.3 | 3615 | UTR 3 | NP_065691.2 | |||
XM_011512912.1 | 3615 | UTR 3 | XP_011511214.1 | |||
XM_011512914.1 | 3615 | UTR 3 | XP_011511216.1 | |||
XM_011512915.2 | 3615 | UTR 3 | XP_011511217.1 | |||
XM_011512916.2 | 3615 | UTR 3 | XP_011511218.1 | |||
XM_017006633.1 | 3615 | UTR 3 | XP_016862122.1 | |||
XM_017006634.1 | 3615 | UTR 3 | XP_016862123.1 | |||
XM_017006635.1 | 3615 | Intron | XP_016862124.1 |