Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGGCTGGGGGAGATGGTGGAGTA[G/T]CTCCAGGCAGACCTACTCACTGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602683 | ||||||||||||||||||||
Literature Links: |
ADGRB2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADGRB2 - adhesion G protein-coupled receptor B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001294335.1 | Intron | NP_001281264.1 | ||||
NM_001294336.1 | Intron | NP_001281265.1 | ||||
XM_011541848.2 | Intron | XP_011540150.1 | ||||
XM_011541849.2 | Intron | XP_011540151.1 | ||||
XM_011541858.2 | Intron | XP_011540160.1 | ||||
XM_017001899.1 | Intron | XP_016857388.1 | ||||
XM_017001900.1 | Intron | XP_016857389.1 | ||||
XM_017001901.1 | Intron | XP_016857390.1 | ||||
XM_017001902.1 | Intron | XP_016857391.1 | ||||
XM_017001903.1 | Intron | XP_016857392.1 | ||||
XM_017001904.1 | Intron | XP_016857393.1 | ||||
XM_017001905.1 | Intron | XP_016857394.1 | ||||
XM_017001906.1 | Intron | XP_016857395.1 | ||||
XM_017001907.1 | Intron | XP_016857396.1 | ||||
XM_017001908.1 | Intron | XP_016857397.1 | ||||
XM_017001909.1 | Intron | XP_016857398.1 | ||||
XM_017001910.1 | Intron | XP_016857399.1 | ||||
XM_017001911.1 | Intron | XP_016857400.1 | ||||
XM_017001912.1 | Intron | XP_016857401.1 |
MIR4254 - microRNA 4254 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |