Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTCTGGTCCCTTACAGCAAAAGT[C/T]TACTGACCCCTGCCCTAAACACAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
68 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612106 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZBTB40 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
ZBTB40 - zinc finger and BTB domain containing 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083621.1 | 162 | UTR 5 | NP_001077090.1 | |||
NM_014870.3 | 162 | Intron | NP_055685.3 | |||
XM_011542499.2 | 162 | Intron | XP_011540801.1 | |||
XM_011542500.2 | 162 | Intron | XP_011540802.1 | |||
XM_017003003.1 | 162 | Intron | XP_016858492.1 |
Set Membership: |
HapMap |