Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
27 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 184755 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ECHDC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ECHDC2 - enoyl-CoA hydratase domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR1273F - microRNA 1273f | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR1273G - microRNA 1273g | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR5095 - microRNA 5095 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCP2 - sterol carrier protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007098.2 | Intron | NP_001007099.1 | ||||
NM_001007099.2 | Intron | NP_001007100.1 | ||||
NM_001007100.2 | Intron | NP_001007101.1 | ||||
NM_001007250.2 | Intron | NP_001007251.1 | ||||
NM_001193599.1 | Intron | NP_001180528.1 | ||||
NM_001193600.1 | Intron | NP_001180529.1 | ||||
NM_001193617.1 | Intron | NP_001180546.1 | ||||
NM_002979.4 | Intron | NP_002970.2 | ||||
XM_005271103.4 | Intron | XP_005271160.1 | ||||
XM_011541935.2 | Intron | XP_011540237.1 | ||||
XM_017002045.1 | Intron | XP_016857534.1 | ||||
XM_017002046.1 | Intron | XP_016857535.1 |
Set Membership: |
HapMap |