Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTCACAAGTATGTACTGAGAGCC[C/T]ATTAGGTGTGAGGGAAATGAAGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
43 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603417 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC148709 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC148709 - actin pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RABIF - RAB interacting factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002871.4 | 2708 | UTR 3 | NP_002862.2 |
Set Membership: |
HapMap |