Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606542 MIM: 612187 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HDAC7 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HDAC7 - histone deacetylase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098416.3 | Intron | NP_001091886.1 | ||||
NM_001308090.1 | Intron | NP_001295019.1 | ||||
NM_015401.4 | Intron | NP_056216.2 | ||||
XM_011538478.1 | Intron | XP_011536780.1 | ||||
XM_011538479.1 | Intron | XP_011536781.1 | ||||
XM_011538480.1 | Intron | XP_011536782.1 | ||||
XM_011538481.1 | Intron | XP_011536783.1 | ||||
XM_011538482.1 | Intron | XP_011536784.1 | ||||
XM_011538483.1 | Intron | XP_011536785.1 | ||||
XM_017019455.1 | Intron | XP_016874944.1 | ||||
XM_017019456.1 | Intron | XP_016874945.1 |
SLC48A1 - solute carrier family 48 member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |