Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCCCAGAGTATATACTGAGATAC[A/G]GAAGGATGATGGCTCATTCATGATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603344 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
USO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
USO1 - USO1 vesicle transport factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290049.1 | Intron | NP_001276978.1 | ||||
NM_003715.3 | Intron | NP_003706.2 | ||||
XM_006714396.3 | Intron | XP_006714459.1 | ||||
XM_006714397.3 | Intron | XP_006714460.1 |
Set Membership: |
HapMap |