Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCATTAGTGACCTTGGGAAAAGCT[A/G]TGTAGAGGTGATGGTGAGAATGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605774 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KLHL2 - kelch like family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161521.1 | Intron | NP_001154993.1 | ||||
NM_001161522.1 | Intron | NP_001154994.1 | ||||
NM_007246.3 | Intron | NP_009177.3 | ||||
XM_011531572.2 | Intron | XP_011529874.1 | ||||
XM_011531575.2 | Intron | XP_011529877.1 | ||||
XM_011531576.2 | Intron | XP_011529878.1 | ||||
XM_017007674.1 | Intron | XP_016863163.1 | ||||
XM_017007675.1 | Intron | XP_016863164.1 | ||||
XM_017007676.1 | Intron | XP_016863165.1 | ||||
XM_017007677.1 | Intron | XP_016863166.1 |
Set Membership: |
HapMap |