Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATTATTGCAAGTGAAAAAACTAA[A/C]GATGCAAATGAAAGGGGTAGCTATA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
21 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614698 MIM: 602869 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COX20 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
COX20 - COX20, cytochrome c oxidase assembly factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001312871.1 | Intron | NP_001299800.1 | ||||
NM_001312872.1 | Intron | NP_001299801.1 | ||||
NM_001312873.1 | Intron | NP_001299802.1 | ||||
NM_001312874.1 | Intron | NP_001299803.1 | ||||
NM_198076.5 | Intron | NP_932342.1 |
HNRNPU - heterogeneous nuclear ribonucleoprotein U | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |