Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGTGAAGAGTTTTGATCCCTCG[C/T]ACATCCTCGAGCCAGGCAGGGCGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607232 MIM: 607234 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRGPRE PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MRGPRE - MAS related GPR family member E | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRGPRG - MAS related GPR family member G | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRGPRG-AS1 - MRGPRG antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |