Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606037 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD96 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CD96 - CD96 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318889.1 | Intron | NP_001305818.1 | ||||
NM_005816.4 | Intron | NP_005807.1 | ||||
NM_198196.2 | Intron | NP_937839.1 | ||||
XM_005247063.3 | Intron | XP_005247120.1 | ||||
XM_006713469.3 | Intron | XP_006713532.1 | ||||
XM_006713470.3 | Intron | XP_006713533.1 | ||||
XM_017005521.1 | Intron | XP_016861010.1 | ||||
XM_017005522.1 | Intron | XP_016861011.1 |
Set Membership: |
HapMap |