Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAAGCTGTACCTAGTAACCTTCC[A/C]AAATTGTTTTTAAATAAAGTTCTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600788 | ||||||||||||||||||||
Literature Links: |
TCEB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TCEB1 - transcription elongation factor B subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204857.1 | 1161 | Intron | NP_001191786.1 | |||
NM_001204858.1 | 1161 | UTR 3 | NP_001191787.1 | |||
NM_001204859.1 | 1161 | UTR 3 | NP_001191788.1 | |||
NM_001204860.1 | 1161 | UTR 3 | NP_001191789.1 | |||
NM_001204861.1 | 1161 | UTR 3 | NP_001191790.1 | |||
NM_001204862.1 | 1161 | Intron | NP_001191791.1 | |||
NM_001204863.1 | 1161 | Intron | NP_001191792.1 | |||
NM_001204864.1 | 1161 | Intron | NP_001191793.1 | |||
NM_005648.3 | 1161 | Intron | NP_005639.1 | |||
XM_011517580.2 | 1161 | UTR 3 | XP_011515882.1 | |||
XM_011517581.2 | 1161 | UTR 3 | XP_011515883.1 |