Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTCTTGGCTTATCTTTGATGATT[A/T]AAAAATTTTGATATATATTTTATCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 608848 | ||||||||||||||||||||
Literature Links: |
FNBP1L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FNBP1L - formin binding protein 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024948.2 | Intron | NP_001020119.1 | ||||
NM_001164473.2 | Intron | NP_001157945.1 | ||||
NM_017737.4 | Intron | NP_060207.2 | ||||
XM_011541625.2 | Intron | XP_011539927.1 | ||||
XM_017001533.1 | Intron | XP_016857022.1 |