Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCCCTTCCCAGCCCTGGGCGTCC[A/G]CCTCCCCCGCCACCTCCCAGGCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602409 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MLLT10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MLLT10 - myeloid/lymphoid or mixed-lineage leukemia; translocated to, 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195626.1 | Intron | NP_001182555.1 | ||||
NM_001195627.1 | Intron | NP_001182556.1 | ||||
NM_001195628.1 | Intron | NP_001182557.1 | ||||
NM_001195630.1 | Intron | NP_001182559.1 | ||||
NM_001324296.1 | Intron | NP_001311225.1 | ||||
NM_001324297.1 | Intron | NP_001311226.1 | ||||
NM_004641.3 | Intron | NP_004632.1 |
SKIDA1 - SKI/DACH domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |