Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGCTGCATGCACCCTGGGGGCT[A/G]TGGGTTGGACAAGCTCGTTATAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HECTD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HECTD2 - HECT domain E3 ubiquitin protein ligase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284274.1 | Intron | NP_001271203.1 | ||||
NM_173497.3 | Intron | NP_775768.3 | ||||
NM_182765.4 | Intron | NP_877497.3 | ||||
XM_017015750.1 | Intron | XP_016871239.1 | ||||
XM_017015751.1 | Intron | XP_016871240.1 | ||||
XM_017015752.1 | Intron | XP_016871241.1 |
HECTD2-AS1 - HECTD2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |