Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATTTTTAATACAAGTTAATGTGT[C/T]GCTGAGCAGTAGGCTCTTTTTGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612352 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
STAMBPL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STAMBPL1 - STAM binding protein like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020799.3 | Intron | NP_065850.1 | ||||
XM_006717928.2 | Intron | XP_006717991.1 | ||||
XM_006717930.3 | Intron | XP_006717993.1 | ||||
XM_011539983.1 | Intron | XP_011538285.1 | ||||
XM_011539985.1 | Intron | XP_011538287.1 | ||||
XM_017016453.1 | Intron | XP_016871942.1 | ||||
XM_017016454.1 | Intron | XP_016871943.1 | ||||
XM_017016455.1 | Intron | XP_016871944.1 | ||||
XM_017016456.1 | Intron | XP_016871945.1 | ||||
XM_017016457.1 | Intron | XP_016871946.1 | ||||
XM_017016458.1 | Intron | XP_016871947.1 |