Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTACGAAAGGTCCTGTTGTAACA[C/G]AAAATCTCTGATAAAACAGATAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613922 MIM: 613924 | ||||||||||||||||||||
Literature Links: |
LIPK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LIPK - lipase family member K | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LIPN - lipase family member N | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102469.1 | Intron | NP_001095939.1 | ||||
XM_005270049.3 | Intron | XP_005270106.1 | ||||
XM_011540083.2 | Intron | XP_011538385.1 | ||||
XM_011540084.2 | Intron | XP_011538386.1 | ||||
XM_011540085.2 | Intron | XP_011538387.1 | ||||
XM_011540086.2 | Intron | XP_011538388.1 | ||||
XM_017016549.1 | Intron | XP_016872038.1 |