Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 136470 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FST PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FST - follistatin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006350.3 | 48 | Intron | NP_006341.1 | |||
NM_013409.2 | 48 | Intron | NP_037541.1 | |||
XM_005248400.4 | 48 | Intron | XP_005248457.1 | |||
XM_005248401.4 | 48 | Intron | XP_005248458.1 | |||
XM_005248402.4 | 48 | Intron | XP_005248459.1 | |||
XM_005248403.4 | 48 | Intron | XP_005248460.1 | |||
XM_011543099.2 | 48 | UTR 5 | XP_011541401.1 | |||
XM_017008954.1 | 48 | Intron | XP_016864443.1 | |||
XM_017008955.1 | 48 | Intron | XP_016864444.1 |
Set Membership: |
HapMap |