Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACTGTGTGGTCAGGGAGGGTTCC[C/T]GAACACCTGGCCTCCCAGGCCAGAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC91450 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC91450 - uncharacterized LOC91450 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBC1D2B - TBC1 domain family member 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015079.5 | Intron | NP_055894.6 | ||||
NM_144572.1 | Intron | NP_653173.1 | ||||
XM_011521387.2 | Intron | XP_011519689.1 | ||||
XM_017022022.1 | Intron | XP_016877511.1 |
Set Membership: |
HapMap |