Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
TAAACCAGCTCGAAGGAGCCTTCCA[A/G]ACTTTGCCCTGCCGAACCCTCTCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601593 | ||||||||||||||||||||
Literature Links: |
BARD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BARD1 - BRCA1 associated RING domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000465.3 | 868 | Silent Mutation | GTC,GTT | V,V 755 | NP_000456.2 | |
NM_001282543.1 | 868 | Silent Mutation | GTC,GTT | V,V 736 | NP_001269472.1 | |
NM_001282545.1 | 868 | Silent Mutation | GTC,GTT | V,V 304 | NP_001269474.1 | |
NM_001282548.1 | 868 | Silent Mutation | GTC,GTT | V,V 285 | NP_001269477.1 | |
NM_001282549.1 | 868 | Silent Mutation | GTC,GTT | V,V 242 | NP_001269478.1 | |
XM_017004613.1 | 868 | Silent Mutation | GTC,GTT | V,V 788 | XP_016860102.1 | |
XM_017004614.1 | 868 | Intron | XP_016860103.1 |