Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGCGAGCACAGCCCATAAGGGTG[C/T]TAAAAGTAGTACATGAATGTGAGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FAM198B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM198B - family with sequence similarity 198 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031700.2 | 3642 | UTR 3 | NP_001026870.2 | |||
NM_001128424.1 | 3642 | UTR 3 | NP_001121896.1 | |||
NM_016613.6 | 3642 | UTR 3 | NP_057697.2 |