Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATTAACTGCTTCTAGGCAATGCC[A/C]CAGCAAACTAATCAGGCAGTGGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600597 | ||||||||||||||||||||
Literature Links: |
PLCL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLCL1 - phospholipase C like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006226.3 | Intron | NP_006217.3 | ||||
XM_005246643.3 | Intron | XP_005246700.1 | ||||
XM_005246644.3 | Intron | XP_005246701.1 | ||||
XM_011511351.2 | Intron | XP_011509653.1 | ||||
XM_017004339.1 | Intron | XP_016859828.1 | ||||
XM_017004340.1 | Intron | XP_016859829.1 |