Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAGTCACTAACCTCCACTCCCAA[C/T]GTTCTTCACAAATTCCTGCCAATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614136 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRAPPC8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TRAPPC8 - trafficking protein particle complex 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014939.3 | Intron | NP_055754.2 | ||||
XM_005258233.2 | Intron | XP_005258290.1 | ||||
XM_005258234.2 | Intron | XP_005258291.1 | ||||
XM_005258235.3 | Intron | XP_005258292.1 | ||||
XM_006722420.3 | Intron | XP_006722483.1 | ||||
XM_011525884.2 | Intron | XP_011524186.1 | ||||
XM_017025615.1 | Intron | XP_016881104.1 | ||||
XM_017025616.1 | Intron | XP_016881105.1 | ||||
XM_017025617.1 | Intron | XP_016881106.1 |