Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCATCCAGCTCCACAGCTCCCAAA[A/G]CCCACGAAGGAAGCTTTGAAATATC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601459 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PNOC PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PNOC - prepronociceptin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284244.1 | Intron | NP_001271173.1 | ||||
NM_006228.4 | Intron | NP_006219.1 | ||||
XM_005273532.2 | Intron | XP_005273589.1 | ||||
XM_011544559.2 | Intron | XP_011542861.1 | ||||
XM_017013578.1 | Intron | XP_016869067.1 |
Set Membership: |
HapMap |