Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGTCTCCAGAAAGCAAATCCCCCA[A/G]CAATCCTGTCACTTGTCCCAGAATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
NUGGC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NUGGC - nuclear GTPase, germinal center associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010906.1 | Intron | NP_001010906.1 | ||||
XM_011544523.2 | Intron | XP_011542825.1 | ||||
XM_011544524.2 | Intron | XP_011542826.1 | ||||
XM_011544525.1 | Intron | XP_011542827.1 | ||||
XM_011544526.2 | Intron | XP_011542828.1 | ||||
XM_017013403.1 | Intron | XP_016868892.1 |