Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615427 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELMOD3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ELMOD3 - ELMO domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135021.1 | Intron | NP_001128493.1 | ||||
NM_001135022.1 | Intron | NP_001128494.1 | ||||
NM_001135023.1 | Intron | NP_001128495.1 | ||||
NM_032213.4 | Intron | NP_115589.2 | ||||
XM_005264596.3 | Intron | XP_005264653.1 | ||||
XM_005264597.3 | Intron | XP_005264654.1 | ||||
XM_005264598.3 | Intron | XP_005264655.1 | ||||
XM_017005078.1 | Intron | XP_016860567.1 | ||||
XM_017005079.1 | Intron | XP_016860568.1 |
RETSAT - retinol saturase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |