Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAAAACCTTATTGCTGTTTTGAT[A/T]CAAAATACAGTATGAAGATACCTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FAM126B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM126B - family with sequence similarity 126 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321618.1 | 1983 | UTR 3 | NP_001308547.1 | |||
NM_001321619.1 | 1983 | UTR 3 | NP_001308548.1 | |||
NM_001321621.1 | 1983 | UTR 3 | NP_001308550.1 | |||
NM_001321622.1 | 1983 | UTR 3 | NP_001308551.1 | |||
NM_001321623.1 | 1983 | UTR 3 | NP_001308552.1 | |||
NM_001321624.1 | 1983 | UTR 3 | NP_001308553.1 | |||
NM_001321625.1 | 1983 | UTR 3 | NP_001308554.1 | |||
NM_001321626.1 | 1983 | UTR 3 | NP_001308555.1 | |||
NM_001321627.1 | 1983 | UTR 3 | NP_001308556.1 | |||
NM_001321628.1 | 1983 | UTR 3 | NP_001308557.1 | |||
NM_001321629.1 | 1983 | UTR 3 | NP_001308558.1 | |||
NM_173822.4 | 1983 | UTR 3 | NP_776183.1 | |||
XM_006712443.3 | 1983 | UTR 3 | XP_006712506.1 | |||
XM_011511002.2 | 1983 | UTR 3 | XP_011509304.1 | |||
XM_017003879.1 | 1983 | UTR 3 | XP_016859368.1 | |||
XM_017003880.1 | 1983 | UTR 3 | XP_016859369.1 | |||
XM_017003881.1 | 1983 | UTR 3 | XP_016859370.1 | |||
XM_017003882.1 | 1983 | UTR 3 | XP_016859371.1 | |||
XM_017003883.1 | 1983 | UTR 3 | XP_016859372.1 |