Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTAAGGCTTCTCTGAGAGTAATT[C/T]TTTTGGCTGGATCATACTCCAACAT
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 601951 | |||||||||||||||||||||||||||||
Literature Links: |
CLK1 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
CLK1 - CDC like kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162407.1 | 1575 | Missense Mutation | AAA,AGA | K,R 507 | NP_001155879.1 | |
NM_004071.3 | 1575 | Missense Mutation | AAA,AGA | K,R 465 | NP_004062.2 |
Set Membership: |
HapMap |