Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTTGATTGTTTTTTATCAGACCA[A/G]TACAAACATGCAGGAAAAAGGGCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615930 MIM: 609590 | ||||||||||||||||||||
Literature Links: |
CAHM PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAHM - colon adenocarcinoma hypermethylated (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
QKI - QKI, KH domain containing RNA binding | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301085.1 | Intron | NP_001288014.1 | ||||
NM_006775.2 | Intron | NP_006766.1 | ||||
NM_206853.2 | Intron | NP_996735.1 | ||||
NM_206854.2 | Intron | NP_996736.1 | ||||
NM_206855.2 | Intron | NP_996737.1 | ||||
XM_011536258.2 | Intron | XP_011534560.1 | ||||
XM_011536259.2 | Intron | XP_011534561.1 | ||||
XM_011536260.2 | Intron | XP_011534562.1 | ||||
XM_011536261.1 | Intron | XP_011534563.1 | ||||
XM_017011502.1 | Intron | XP_016866991.1 | ||||
XM_017011503.1 | Intron | XP_016866992.1 | ||||
XM_017011504.1 | Intron | XP_016866993.1 |