Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTACCCACTAGGACATTCACATTC[A/C]TGTGCTGGTGAAAACGGCTTTGTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 125255 | ||||||||||||||||||||
Literature Links: |
DCN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DCN - decorin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001920.4 | Intron | NP_001911.1 | ||||
NM_133503.3 | Intron | NP_598010.1 | ||||
NM_133504.3 | Intron | NP_598011.1 | ||||
NM_133505.3 | Intron | NP_598012.1 | ||||
NM_133506.3 | Intron | NP_598013.1 | ||||
NM_133507.3 | Intron | NP_598014.1 | ||||
XM_005268693.1 | Intron | XP_005268750.1 | ||||
XM_006719270.1 | Intron | XP_006719333.1 | ||||
XM_017018917.1 | Intron | XP_016874406.1 |