Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTTATCCCTTCTGTCTGAGTCTA[A/G]GATAGGGGATTACTAATGGTGGCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 608661 | ||||||||||||||||||||
Literature Links: |
PODN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PODN - podocan | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153703.4 | Intron | NP_714914.2 | ||||
XM_005270457.3 | Intron | XP_005270514.1 | ||||
XM_011540677.2 | Intron | XP_011538979.1 | ||||
XM_017000294.1 | Intron | XP_016855783.1 |