Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTGAAGGGCCACTCCAAAGGTT[A/C]TAAAGGGATTGGAAATGTATGCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600698 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMGA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HMGA2 - high mobility group AT-hook 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300918.1 | Intron | NP_001287847.1 | ||||
NM_001300919.1 | Intron | NP_001287848.1 | ||||
NM_003483.4 | Intron | NP_003474.1 | ||||
NM_003484.1 | Intron | NP_003475.1 | ||||
XM_017019989.1 | Intron | XP_016875478.1 |
RPSAP52 - ribosomal protein SA pseudogene 52 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |