Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAGAAACACTGCCTAGGGTGTGT[A/T]TTGCTGATGTTGCTGGGCCGGTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604207 MIM: 611871 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPNE3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CPNE3 - copine 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003909.4 | Intron | NP_003900.1 | ||||
XM_005251093.4 | Intron | XP_005251150.1 | ||||
XM_017013945.1 | Intron | XP_016869434.1 |
RMDN1 - regulator of microtubule dynamics 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |