Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAAATTTAATTTTGTAGTTTGC[C/T]CCTTACCTTTACGACCTTTAAATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602307 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WWP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WWP1 - WW domain containing E3 ubiquitin protein ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007013.3 | Intron | NP_008944.1 | ||||
XM_005250760.3 | Intron | XP_005250817.1 | ||||
XM_005250761.3 | Intron | XP_005250818.1 | ||||
XM_011516802.2 | Intron | XP_011515104.1 | ||||
XM_017012993.1 | Intron | XP_016868482.1 | ||||
XM_017012994.1 | Intron | XP_016868483.1 | ||||
XM_017012995.1 | Intron | XP_016868484.1 | ||||
XM_017012996.1 | Intron | XP_016868485.1 | ||||
XM_017012997.1 | Intron | XP_016868486.1 |