Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRELID2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PRELID2 - PRELI domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138492.5 | Intron | NP_612501.3 | ||||
NM_182960.3 | Intron | NP_892005.1 | ||||
NM_205846.2 | Intron | NP_995318.1 | ||||
XM_017009127.1 | Intron | XP_016864616.1 | ||||
XM_017009128.1 | Intron | XP_016864617.1 | ||||
XM_017009129.1 | Intron | XP_016864618.1 | ||||
XM_017009130.1 | Intron | XP_016864619.1 | ||||
XM_017009131.1 | Intron | XP_016864620.1 | ||||
XM_017009132.1 | Intron | XP_016864621.1 | ||||
XM_017009133.1 | Intron | XP_016864622.1 | ||||
XM_017009134.1 | Intron | XP_016864623.1 | ||||
XM_017009135.1 | Intron | XP_016864624.1 |